Learn More
Leica Biosystems SLIDE WHT XADH 1440/CASE
Catalog No. NC2293477
Encompass
X-tra Slides have a positive charged surface which helps to bond tissue sections and cytology preparations. X‐tra slides are used for demanding H&E procedures that require tissue adhesion. Size: 1” x 3” x .04” (unless otherwise noted)Hydrophobic surface with approximately 90 degree contact angle. Recommended uses include special stains, IHC, ISH, cryosectioning, and cytology.
This item is not returnable. View return policy
Catalog No. NC2293477 Supplier Leica Biosystems Supplier No. 3800050
X-tra Slides have a positive charged surface which helps to bond tissue sections and cytology preparations. X‐tra slides are used for demanding H&E procedures that require tissue adhesion. Size: 1” x 3” x .04” (unless otherwise noted)Hydrophobic surface with approximately 90 degree contact angle. Recommended uses include special stains, IHC, ISH, cryosectioning, and cytology.