Learn More
Leica Biosystems SLIDE WHTE SNOWCT CORN CS/1440
Catalog No. NC0318073
Encompass_Preferred
Snowcoat Clipped Corner Slides are clipped at 45° angles to help reduce glass breakage and fragmentation when loaded into slide printers. These slides have been optimized for use with the Leica printer.
Catalog No. NC0318073 Supplier Leica Biosystems Supplier No. 3800381
Snowcoat Clipped Corner Slides are clipped at 45° angles to help reduce glass breakage and fragmentation when loaded into slide printers. These slides have been optimized for use with the Leica printer.