Learn More
Med Vet International McKesson Glass Microscope Slide, 25 x 75 x 1 mm, 70-109PMCK, 72/Box
Catalog No. 503043881
Encompass_Preferred
Microscope Slide McKesson 25 X 75 X 1 mm Extra-Frosted End 25 x 75 x 1 millimeters White on one end Ground edges Made from the finest quality sheet glass Guaranteed precleaned Non-corrosive glass
Catalog No. 50-304-3881 Supplier Med Vet International Supplier No. 938363BX
Microscope Slide McKesson 25 X 75 X 1 mm Extra-Frosted End 25 x 75 x 1 millimeters White on one end Ground edges Made from the finest quality sheet glass Guaranteed precleaned Non-corrosive glass