Learn More
Med Vet International McKesson Glass Microscope Slide, 1 x 3 Inch x 1 mm, 16-7133, 1440/Case
Catalog No. 503045438
Encompass_Preferred
Microscope Slide McKesson 1 X 3 Inch X 1 mm Extra-Frosted End Extra-frosted 3 inches x 1 inch x 1 millimeter Economy slides designed to meet the needs of a variety of applications Ground edges
Catalog No. 50-304-5438 Supplier Med Vet International Supplier No. 464499CS
Microscope Slide McKesson 1 X 3 Inch X 1 mm Extra-Frosted End Extra-frosted 3 inches x 1 inch x 1 millimeter Economy slides designed to meet the needs of a variety of applications Ground edges