Learn More
Electron Microscopy Sciences The safest choice for a xylene alternative available today. It performs equal-to or better than Xylene, yet the non-hazardous clearing agent is also non-flammable and odorless, and contains no vapor disguising additives.
SDP
Catalog No. NC2824213
Encompass_Preferred
SlideBrite™ is a high quality performance clearing agent that reduces your risks and is the safest xylene alternative available today. It performs equal-to or better than Xylene, yet the non-hazardous clearing agent is also non-flammable and odorless, and contains no vapor disguising additives. This xylene substitute enhances nuclear detail and features a fast slide drying time. It is gentle, yet very effective at deparaffinization. It will not result in any tissue shrinkage or morphology change and there will be no brittle tissue subsequent to tissue processing.
Catalog No. NC2824213 Supplier Electron Microscopy Sciences Supplier No. 2340101
SlideBrite™ is a high quality performance clearing agent that reduces your risks and is the safest xylene alternative available today. It performs equal-to or better than Xylene, yet the non-hazardous clearing agent is also non-flammable and odorless, and contains no vapor disguising additives. This xylene substitute enhances nuclear detail and features a fast slide drying time. It is gentle, yet very effective at deparaffinization. It will not result in any tissue shrinkage or morphology change and there will be no brittle tissue subsequent to tissue processing.