Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ SlideMate™ Laser Plus Microscope Slides

Catalog No. p-200007426 Shop All Epredia Products
Click to view available options
Color:
Blue
Green
Pink
White
Yellow
This item is not returnable. View return policy

Catalog No. 22-042-958

Supplier: Epredia™ LS4041IPS85231

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Optimized for use on the SlideMate Laser Printer, these slides provide for a clear and robust printing surface. The adhesion properties of SlideMate Laser Plus slides are the same technology as Superfrost PLUS™ slides.

  • Epredia SlideMate laser slides are made from extra-white glass, manufactured to high quality standards.
  • The clarity achieved during the manufacturing process helps ensure high levels of transparency to aid in accurate diagnoses.
  • The adhesive properties of the SlideMate Laser Plus Slides makes them suitable for all routine clinical applications.
  • The slides come pre-cleaned, flat with finely ground edges.
  • Each package comes wrapped to help protect against humidity and moisture.
  • The packaging is also designed to assist with loading the slides into SlideMate laser printer hoppers to prevent contamination due to handling.
  • Manufactured in the USA.
  • SlideMate laser slides are smooth, three-quarter inch white, blue, green, pink or yellow tab slides validated to work with the SlideMate laser slide printer.
  • The tab is impervious to virtually all common laboratory chemicals and reagents.

Specifications

Length (Metric) 75 mm
Frosted Not Frosted
Precleaned Yes
For Use With (Equipment) Epredia SlideMate Laser Printer
Edges Finely Ground
Material High Quality Glass
Type Microscope Slide
No. per Case 1440/Cs.
Width (Metric) 25 mm
No. per Pack 144 Pk.
Thickness (Metric) 1 mm
Color White
Coating Adhesive
Corner Style 90 degrees
Length (English) 3 in.
Width (English) 1 in.
Dimensions (L x W) 75 x 25 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.