Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ SlideMate™ Microscope Slides

Catalog No. 22042937 Shop All Epredia Products
Change view
Click to view available options
Color:
Blue
Green
Pink
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-042-937 White
22-042-939 Blue
22-042-948 Pink
22-042-950 Yellow
22-042-952 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 22-042-937 Supplier Epredia™ Supplier No. TT4041I8217001
Add to Cart
Add to Cart
This item is not returnable. View return policy

SlideMate™ Microscope Slides offer crisp, clean printing. Designed for use on SlideMate thermal printers. Reduces identification issues common to handwritten labels when paired with SlideMate printers. Comes 144 slides per gross; 10 gross per case.

  • Manufactured in the USA of Swiss glass; SlideMate slides are smooth, three-quarter inch white tab slides validated to work with the SlideMate AS slide printer.
  • The tab is impervious to virtually all common laboratory chemicals and reagents.
  • Available in adhesion and non-adhesion formulas. SlideMate Plus Adhesion Slides feature adhesion coating for routine clinical applications.
  • Epredia SlideMate slides are made from extra-white glass, manufactured to high quality standards. The clarity achieved during the manufacturing process helps ensure high levels of transparency to aid in accurate diagnoses.
  • SlideMate slides come pre-cleaned, flat with finely ground edges. Each package comes wrapped to help protect against humidity and moisture. The packaging is also designed to assist with loading the slides into SlideMate printer hoppers to prevent contamination due to handling.

Specifications

Length (Metric) 75 mm
No. per Case 1440
Thickness (Metric) 1 mm
Edges Ground
Material Swiss Glass
Width (Metric) 25 mm
For Use With (Equipment) SlideMate printers
No. per Pack 144
Color White
Coating Non-adhesion
Corner Style 90°
Dimensions (L x W) 75 x 25 mm
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.