Learn More
Epredia™ SlideMate™ TSP Slide Printer
Catalog No. 21402344 Shop All Epredia Products
Change view
Click to view available options
Slide Capacity:
144 Slides
Catalog No. Slide Capacity
21-402-344 144 Slides
1 options
Catalog No. 21-402-344 Supplier Epredia™ Supplier No. 8000066
Add to Cart
Add to Cart
Using elevated printing technology, compatible with a wide range of slide types.
  • High-quality direct-to-slide printing helps protect specimens while minimizing traceability errors associated with label transfer
  • Lateral slide movement to help minimize chipped slides
  • Enclosed slide hoppers help reduce contamination from laboratory dust and debris
  • Backed by proven expertise in slide printing technology and support
  • Provides highly reliable microscope slide printing
  • Validated for use with SlideMate™, SlideMate™ PLUS, SlideMate™ Laser, and SlideMate™ Laser PLUS slides
  • Reliable service and support from industry experts
  • Practical slide printing for the modern laboratory
  • Compatible with a wide range of slide types
  • Thermal transfer technology allows printing on a wide selection of slide types
  • LCD with variable pitch adjustment for enhanced ergonomics
  • Small footprint enables placement at the microtome for on-demand printing
Specifications
Product Type Slide Printer
Slide Capacity 144 Slides
Voltage 24 V