Learn More
Micronova SlideMop™ Low-Touch Flat Head Microfiber Mop Cover
SDP
Catalog No. 19999879 Shop All Micronova Manufacturing Products
Change view
Click to view available options
Autoclavable:
Autoclavable
Catalog No. Autoclavable
19-999-879 Autoclavable
1 options
Catalog No. 19-999-879 Supplier Micronova Supplier No. MZCRAA316
Add to Cart
Add to Cart
The SlideMop™ Low-Touch Flat Head Mop cover is a simple, effective mop system featuring sturdy, collapsible plates that easily slide into the microfiber mop cover and lock into place with an effortless downward push.
  • Microfiber mop covers for use with the the Micronova SlideMop™ Flat Head Mop. (For SlideMop frame, see QSM16.)
  • Microfiber mop covers with polyester pad interiors for disinfectant dispersal and particle pick-up
  • Manufactured for use in Controlled Environments, ISO 5 thru 9
  • Good chemical resistance
Specifications
Product Type Mop Cover
Color Code Blue
Sterility Non-sterile
Autoclavable Autoclavable
Disposable Reusable
Color-coded No
Material Synthetic Microfiber, Polyester