Learn More
Research Products International Corp Slide-Out Freezer Rack for 2"" H Boxes, 16 Box Capacity, 4x4 Array, 22 x 5 1/2 x 9 3/4H
SDP
Catalog No. 501367710
Encompass
Ideal freezer racks feature slide out trays that maximize storage space in upright freezers or refrigerators. Constructed of corrosion resistant stainless steel for rugged durability. Racks will accommodate all 5" square x 2" high cardboard or plastic boxes. Racks are available in several convenient sizes and capacities.
Catalog No. 50-136-7710 Supplier Research Products International Corp Supplier No. 247473
Ideal freezer racks feature slide out trays that maximize storage space in upright freezers or refrigerators. Constructed of corrosion resistant stainless steel for rugged durability. Racks will accommodate all 5" square x 2" high cardboard or plastic boxes. Racks are available in several convenient sizes and capacities.