Learn More
Sigma Aldrich Fine Chemicals Biosciences BRAND(R) cavity slides L x W 76 mm x 26 mm, thickness 1.2-1.5 mm, wells 2 | 50EA
Catalog No. NC2241984
Encompass
BRAND(R) cavity slides L x W 76 mm x 26 mm, thickness 1.2-1.5 mm, wells 2 | 50EA
Catalog No. NC2241984 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. BR47553550EA
BRAND(R) cavity slides L x W 76 mm x 26 mm, thickness 1.2-1.5 mm, wells 2 | 50EA