Learn More
Med Vet International Dynarex Microscope Slides, Frosted, corner grounded, 3" x 1" x 1 mm, 1440/Case
Catalog No. 503044805
Encompass_Preferred
Corrosion resistant, precleaned, lint-free and ready to use.Suitable for all medical applications at economical prices. Made of premium, quality sheet glass with ground corners Provides consistent results for all laboratory tests Frosted slide coating retains most laboratory marking methods Writing surface eliminates the need for paper labels and relabeling of slides
Catalog No. 50-304-4805 Supplier Med Vet International Supplier No. 4171
Corrosion resistant, precleaned, lint-free and ready to use.Suitable for all medical applications at economical prices. Made of premium, quality sheet glass with ground corners Provides consistent results for all laboratory tests Frosted slide coating retains most laboratory marking methods Writing surface eliminates the need for paper labels and relabeling of slides