Learn More
Electron Microscopy Sciences Gold Seal Rite-On™ Slides Frosted End Cytogenetics, 3 x 1", 1 mm thick
SDP
Catalog No. 5030157
Encompass_Preferred
GOLDSEAL slides are made from pure white glass (sodalime) and at accurately cut sizes. To preserve the highest quality from manufacturer to ultimate user, all GOLD SEAL slides are hermetically sealed in our Moisture-Barrier system, providing a dry-slide environments during shipment and storage.
Size: 3 x 1"
Thickness: 1 mm
Catalog No. 50-301-57 Supplier Electron Microscopy Sciences Supplier No. 6372205
GOLDSEAL slides are made from pure white glass (sodalime) and at accurately cut sizes. To preserve the highest quality from manufacturer to ultimate user, all GOLD SEAL slides are hermetically sealed in our Moisture-Barrier system, providing a dry-slide environments during shipment and storage.
Size: 3 x 1"
Thickness: 1 mm