Learn More
Electron Microscopy Sciences Gold Seal UltraFrost Frosted End Slides with Patient Info, 3 x 1", 1 mm thick
SDP
Supplier:  Electron Microscopy Sciences 6372905
Encompass
GOLDSEAL slides are made from pure white glass (sodalime) and at accurately cut sizes. To preserve the highest quality from manufacturer to ultimate user, all GOLD SEAL slides are hermetically sealed in our Moisture-Barrier system, providing a dry-slide environments during shipment and storage. Slide has Gold Seal logo.
Size: 3 x 1"
Thickness: 1 mm
Catalog No. 50-301-66