Learn More
Sigma Aldrich Fine Chemicals Biosciences Slides microscope plain si
Catalog No. 502901732
Encompass
Sealed in a moisture absorbing desiccant pack which can function as a drying stand and a storage box. Slides are 1 mm thick.Our microscope slides are made of high-quality white glass with low iron content no green tint with fine texture and transparency these slides are also corrosion-resistant. The edges are ground smooth to help eliminate sharp cutting surfaces and safe handling. They are packed in a special thermoform plastic box to minimize exposure to moisture and dust (each slide is precleaned).Conform to federal specification NNN-S-450.
Catalog No. 50-290-1732 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S89021PAK
Sealed in a moisture absorbing desiccant pack which can function as a drying stand and a storage box. Slides are 1 mm thick.Our microscope slides are made of high-quality white glass with low iron content no green tint with fine texture and transparency these slides are also corrosion-resistant. The edges are ground smooth to help eliminate sharp cutting surfaces and safe handling. They are packed in a special thermoform plastic box to minimize exposure to moisture and dust (each slide is precleaned).Conform to federal specification NNN-S-450.