Learn More
MTC Bio Microscope Slides, 25 x 75mm, 72/PK
Supplier:  MTC Bio M710045
Encompass
This item has a minimum qty of 6 per supplier requirements.
Microscope slides, positive charged, 45 corners, frosted on one end, 25 x 75mm
Catalog No. 50-187-1157