Learn More
StatLab™ SlideWrite Graphite Lab Pencils
SDP
Catalog No. STLSL8008
Change view
Click to view available options
Quantity:
12 Pencils/Pk.
Catalog No. Quantity
STLSL8008 12 Pencils/Pk.
1 options
Catalog No. STLSL8008 Supplier StatLab™ Supplier No. SL8008
Add to Cart
Add to Cart
Soft-lead pencils designed specifically for labeling histology cassettes and slides.
  • Compatible with both histology cassettes and microscope slides
Specifications
Product Type Colored Pencil
Tip Style Soft Tip
Color Black
Autoclavable Not Autoclavable
Sterility Non-sterile
Disposable Reusable
Latex Free Yes
Quantity 12 Pencils/Pk.