Learn More
Grainger BESSEY Bar Clamp, Light Duty, Sliding T Handle, 8 in. Capacity, 1 ea.
Catalog No. 502787919
Encompass_Preferred
This light-duty bar clamp is designed for reliable performance across a variety of industrial and workshop applications, including assembly, gluing, welding, and metalworking. Featuring a sturdy steel construction and a sliding T-handle mechanism, it ensures efficient and secure clamping for professional projects.
  • 8 in maximum jaw opening for versatile project sizes
  • 880 lb clamping force for secure hold
  • 4 in throat depth
  • Sliding T-handle with bar slide design for rapid positioning
  • Spatter-resistant spindle finish suitable for welding tasks
  • Standard swivel pads accommodate angled surfaces
Catalog No. 50-278-7919 Supplier Grainger Supplier No. 19C716
This light-duty bar clamp is designed for reliable performance across a variety of industrial and workshop applications, including assembly, gluing, welding, and metalworking. Featuring a sturdy steel construction and a sliding T-handle mechanism, it ensures efficient and secure clamping for professional projects.
  • 8 in maximum jaw opening for versatile project sizes
  • 880 lb clamping force for secure hold
  • 4 in throat depth
  • Sliding T-handle with bar slide design for rapid positioning
  • Spatter-resistant spindle finish suitable for welding tasks
  • Standard swivel pads accommodate angled surfaces