Learn More
Grainger Bessey Bar Clamp, Extra Heavy Duty, 12 in. Opening, 8 in. Throat Depth, 1 ea.
Catalog No. 502785663
Encompass_Preferred
These all-steel bar clamps are designed for extra heavy-duty industrial applications, providing significant clamping force and durability. The construction features a single-piece, cold-drawn steel rail and a tempered, drop-forged sliding arm to ensure stability and even force distribution. Engineered to perform the work of traditional C-clamps with increased efficiency, this tool series is suitable for demanding industrial tasks.
  • Clamping capacity of 12 in. and throat depth of 8 in.
  • Maximum clamping force of 7,770 lbs.
  • Integrated wedge locks the sliding arm in place on the clamp rail for repetitive tasks.
  • Heat-treated, high-carbon steel spindle resists weld spatter.
  • Swivel MorPad reduces friction and optimizes clamping force at the contact point.
  • Tilting pad design allows for up to 35° of adjustment.
  • Electrogalvanized finish provides corrosion resistance.
Catalog No. 50-278-5663 Supplier Grainger Supplier No. 19C710
These all-steel bar clamps are designed for extra heavy-duty industrial applications, providing significant clamping force and durability. The construction features a single-piece, cold-drawn steel rail and a tempered, drop-forged sliding arm to ensure stability and even force distribution. Engineered to perform the work of traditional C-clamps with increased efficiency, this tool series is suitable for demanding industrial tasks.
  • Clamping capacity of 12 in. and throat depth of 8 in.
  • Maximum clamping force of 7,770 lbs.
  • Integrated wedge locks the sliding arm in place on the clamp rail for repetitive tasks.
  • Heat-treated, high-carbon steel spindle resists weld spatter.
  • Swivel MorPad reduces friction and optimizes clamping force at the contact point.
  • Tilting pad design allows for up to 35° of adjustment.
  • Electrogalvanized finish provides corrosion resistance.