Learn More
Grainger Sliding Closet Door Floor Guide HDPE PK4
Catalog No. 502765308
Encompass_Preferred
The PRIME-LINE Sliding Closet Door Guide is a floor-mounted guide designed for use with sliding closet doors. It is made from steel with a white powder coat finish and measures 4 inches tall. This guide features nylon rollers and can support doors up to 1-3/8 inches thick and weighing up to 75 pounds per panel. It allows the door to slide smoothly and reduces noise. The guide comes with mounting hardware and installation instructions. It is durable, easy to install, and provides quiet operation for sliding closet doors.
Catalog No. 50-276-5308 Supplier Grainger Supplier No. 170A70
The PRIME-LINE Sliding Closet Door Guide is a floor-mounted guide designed for use with sliding closet doors. It is made from steel with a white powder coat finish and measures 4 inches tall. This guide features nylon rollers and can support doors up to 1-3/8 inches thick and weighing up to 75 pounds per panel. It allows the door to slide smoothly and reduces noise. The guide comes with mounting hardware and installation instructions. It is durable, easy to install, and provides quiet operation for sliding closet doors.