Learn More
Grainger Sliding Closet Door Guide Screws PK4
Catalog No. 502765451
Encompass_Preferred
The PRIME-LINE Sliding Closet Door Guide is a floor-mounted guide for sliding closet doors. It is made of steel with a white powder coat finish and measures 4 inches tall. The guide has nylon rollers and can support doors up to 1-3/8 inches thick and weighing up to 75 lbs per panel. It allows doors to slide smoothly and reduces noise. The Prime-Line sliding door guide is easy to install with included hardware and instructions. It is a high quality closet door guide at an affordable price.
Catalog No. 50-276-5451 Supplier Grainger Supplier No. 170A66
The PRIME-LINE Sliding Closet Door Guide is a floor-mounted guide for sliding closet doors. It is made of steel with a white powder coat finish and measures 4 inches tall. The guide has nylon rollers and can support doors up to 1-3/8 inches thick and weighing up to 75 lbs per panel. It allows doors to slide smoothly and reduces noise. The Prime-Line sliding door guide is easy to install with included hardware and instructions. It is a high quality closet door guide at an affordable price.