Learn More
Ampco Sliding T-Bar with 1/4 in. Drive
SDP
Catalog No. 19174601 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
4 -1/2 in.
Catalog No. Length (English)
19-174-601 4 -1/2 in.
1 options
Catalog No. 19-174-601 Supplier Ampco Supplier No. T1/4
Add to Cart
Add to Cart
Used to drive sockets for tightening and loosening fasteners and to offer adjustable leverage.
  • Non-sparking, non-magnetic, and corrosion-resistant.
  • 4 and 1/2 in. length
Specifications
Product Type Sliding T-Bar
Length (English) 4 -1/2 in.
Includes Sliding T-Bar with 1/4 in. Drive Size
Quantity 1 Ea.