Learn More
Ampco Sliding T-Bar with 3/4 in. Drive
SDP
Catalog No. 19174604 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
18 in.
Catalog No. Length (English)
19-174-604 18 in.
1 options
Catalog No. 19-174-604 Supplier Ampco Supplier No. T3/4
Add to Cart
Add to Cart
Used to drive sockets for tightening and loosening fasteners and to offer adjustable leverage.
  • Non-sparking, non-magnetic, and corrosion-resistant.
  • 18 in. length
Specifications
Product Type Sliding T-Bar
Length (English) 18 in.
Includes Sliding T-Bar with 3/4 in. Drive Size
Quantity 1 Ea.