Learn More
Grainger Sliding Window Sash Locks Black PK2
Catalog No. 502765458
Encompass_Preferred
The PRIME-LINE Sliding Window Sash Locks are high-security locks designed for sliding windows. They feature a dual-locking mechanism with an adjustable hook that engages the window track and a spring-loaded plunger that fits into a hole in the window frame. The locks are constructed from zinc alloy with a chrome finish and measure 3-1/4" long x 1-1/8" wide x 1/2" thick. They come with screws for easy installation and provide an economical way to add security to sliding windows. The sash locks are available in packs of 1, 2, 4, or 12.
Catalog No. 50-276-5458 Supplier Grainger Supplier No. 170D79
The PRIME-LINE Sliding Window Sash Locks are high-security locks designed for sliding windows. They feature a dual-locking mechanism with an adjustable hook that engages the window track and a spring-loaded plunger that fits into a hole in the window frame. The locks are constructed from zinc alloy with a chrome finish and measure 3-1/4" long x 1-1/8" wide x 1/2" thick. They come with screws for easy installation and provide an economical way to add security to sliding windows. The sash locks are available in packs of 1, 2, 4, or 12.