Learn More
Grainger Sliding Window Sash Locks White PK2
Catalog No. 502765314
Encompass_Preferred
The PRIME-LINE Sliding Window Sash Locks are high-security locks designed for sliding windows. They feature a dual-locking mechanism with an adjustable cam that allows the window to be locked partially open. The locks are constructed from zinc alloy with a satin nickel finish and measure 3-1/4" long x 1-1/8" wide x 1/2" high. They come with mounting hardware and installation instructions. The locks help prevent unwanted entry through sliding windows while still allowing windows to be opened for ventilation.
Catalog No. 50-276-5314 Supplier Grainger Supplier No. 170D80
The PRIME-LINE Sliding Window Sash Locks are high-security locks designed for sliding windows. They feature a dual-locking mechanism with an adjustable cam that allows the window to be locked partially open. The locks are constructed from zinc alloy with a satin nickel finish and measure 3-1/4" long x 1-1/8" wide x 1/2" high. They come with mounting hardware and installation instructions. The locks help prevent unwanted entry through sliding windows while still allowing windows to be opened for ventilation.