Learn More
Grainger Sliding Window Tilt Latch Mortise White
Catalog No. 502765465
Encompass_Preferred
The PRIME-LINE Sliding Window Tilt Latch is a high-quality tilt latch designed for sliding windows. It is constructed from steel with a zinc plated finish and features an adjustable keeper that allows for a custom fit. This tilt latch has a low profile design with a flat front plate that sits flush against the window frame. It comes with all necessary mounting hardware and has a locking mechanism to securely hold the window in a tilted open position. This product provides an easy way to add ventilation control to sliding windows. With its durable construction, it is built to last while blending in discreetly.
Catalog No. 50-276-5465 Supplier Grainger Supplier No. 170D91
The PRIME-LINE Sliding Window Tilt Latch is a high-quality tilt latch designed for sliding windows. It is constructed from steel with a zinc plated finish and features an adjustable keeper that allows for a custom fit. This tilt latch has a low profile design with a flat front plate that sits flush against the window frame. It comes with all necessary mounting hardware and has a locking mechanism to securely hold the window in a tilted open position. This product provides an easy way to add ventilation control to sliding windows. With its durable construction, it is built to last while blending in discreetly.