Learn More
Cayman Chemical SLIGRLNH2trIfluoroacetAT 10mg
SDP
Catalog No. 502966242
Encompass_Preferred
A recombinant peptide that activates PAR-2 (EC50= ~5 µM), without requiring receptor cleavage; used to explore signaling through PAR2 in cells and in animals
Catalog No. 50-296-6242 Supplier Cayman Chemical Supplier No. 1672310 MG
A recombinant peptide that activates PAR-2 (EC50= ~5 µM), without requiring receptor cleavage; used to explore signaling through PAR2 in cells and in animals