Learn More
Electron Microscopy Sciences B.O.P. Biopsy Organizer Plate Slim Light Pad with Adapter
SDP
Catalog No. 5033451
Encompass
This light box is ultra thin, only 1⁄2" (14mm) thick. This new technology provides bright, even 5400K light for illuminating specimen containers and easily finding even small specimen material with the clear plastic.
Catalog No. 50-334-51 Supplier Electron Microscopy Sciences Supplier No. 6392060
This light box is ultra thin, only 1⁄2" (14mm) thick. This new technology provides bright, even 5400K light for illuminating specimen containers and easily finding even small specimen material with the clear plastic.