Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Micronova™ SlimLine™ Cleanroom Cart System
SDP

Catalog No. 191304847 Shop All Micronova Manufacturing Products
Change view
Click to view available options
Product Type:
Triple Bucket Cart
Color:
Blue, Yellow, Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Color
19-130-4847 Triple Bucket Cart Blue, Yellow, Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-130-4847 Supplier Micronova™ Supplier No. C33S

missing translation for 'validMainframeMsg'

Includes

Each system supplied with 6-gal. (22.7L) polypropylene buckets. Two-bucket system includes one blue and one yellow bucket; three-bucket system includes one blue, one red, and one yellow bucket. Optional wringer, basket, bucket liner and replacement buckets sold separately.

A durable, double or triple bucket cart system with an electropolished, stainless steel frame, optional wringer bar, and three grip-it holders for easy storage and interchange of handles and mop heads. Systems comes standard with color coded, 22.7 liter (6 gal.) polypropylene buckets.

The Micronova SlimLine™ Stainless Steel Cart Systems include an electropolished stainless steel cart and come standard with either two or three, color-coded, polypropylene buckets to distinguish between clean and rinse buckets. The three grip-it holders at the back of the cart allow for easy interchange and storage of handles and mop heads. An optional wringer bar accommodates either the Downpress or Flat Head Wringers, allowing placement above the rinse bucket for simplified wringing.

  • Cart is crafted from electropolished stainless steel for durability and long life
  • Includes three grip-it holders at the back of the cart
  • Easy storage and interchange of handles and mop heads
  • Comes standard with color-coded, 6-gallon (22-L), polypropylene buckets
  • Double bucket system comes with 1 blue and 1 yellow bucket
  • Triple bucket system includes an additional red bucket
  • Optional wringer bar allows wringers to be placed above the rinse bucket
  • Buckets are best autoclaved in the inverted position
  • Entire unit is autoclavable

Specifications

Autoclavable Autoclavable
Product Type Triple Bucket Cart
Capacity (English) 18 gal. (6 gal./Bucket)
Capacity (Metric) 68.1 L (22.7 L/bucket)
Color Blue, Yellow, Red
For Use With (Equipment) SlimLine™ Cleanroom Cart System
Includes 3 Buckets
Material Stainless Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.