Learn More
StatLab Slimline Slide Drying Hotplate
SDP
Catalog No. 21010361
Change view
Click to view available options
Slide Capacity:
12
Catalog No. Slide Capacity
21-010-361 12
1 options
This item is not returnable. View return policy
Catalog No. 21-010-361 Supplier StatLab Supplier No. SISDHSLIM
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed to efficiently dry slides through the use of heated air radiating from a hotplate beneath the slides.
  • An ultra-fast heating system ensures rapid warm up, and digital controls provide temperature accuracy
  • Microscope slides are supported on four movable rails which allow heated air to pass beneath the slides to accelerate drying, while slides sit at a sloped angle to encourage water drain off
  • The rails can be removed from the instrument so that slides can be placed directly on the heated surface when required
  • An optional stainless steel lid (sold separately, SI-SDH-LID) is available which can be used to further reduce drying times and efficiency
  • Features: Variable digital temperature control, ultra-fast heating system for rapid warm up, movable rails, anti-microbial powder coat finish, memory function for desired temperature at start-up, integrated circuit breakers
  • Dimensions: 183 x 450 x 85.5 mm (D X W X H)
  • Weight: 4.4 lb. (2.2 Kg)
Specifications
Product Type Hotplate
Slide Capacity 12