Learn More
StatLab Slimsette Embedding Cassettes
SDP
Catalog No. 21033438
Change view
Click to view available options
Color:
Blue
Green
Lilac
Peach
Pink
Tan
White
Yellow
Pore Style:
Bulk
Sleeved
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Pore Style
21-033-438 Tan Sleeved
21-033-427 Lilac Sleeved
21-033-428 White Bulk
21-033-429 White Sleeved
21-033-430 Pink Sleeved
21-033-431 Green Bulk
21-033-432 Green Sleeved
21-033-433 Yellow Bulk
21-033-434 Yellow Sleeved
21-033-435 Blue Bulk
21-033-436 Blue Sleeved
21-033-437 Peach Sleeved
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 21-033-438 Supplier StatLab Supplier No. M5098SL
Add to Cart
Add to Cart
Designed to be more compact than standard cassettes
  • Design includes a recessed cover that allows more cassettes to be stacked in automatic labelers, processing baskets and storage cabinets
  • 45-Degree labeling surface, lids attached
  • Sleeved, CS750
  • Bulk packed, CS1500
Specifications
Pore Style Sleeved
Color Tan
Product Type Embedding Cassette