Learn More
ECM Bioscience Slingshot-1L Phospho-Regulation Antibody Sampler Kit, ECM Biosciences
Catalog No. 502068836
Encompass_Preferred
The Slingshot-1L (SSH1L) phospho-regulation antibody sampler kit can be used to detect the level of phosphorylation of SSH1L (Ser-978) relative to the total expression level of SSH1L. The kit also includes peptides for antibody blocking experiments and a secondary reagent for antibody detection. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms
Catalog No. 50-206-8836 Supplier ECM Bioscience Supplier No. SK6410
The Slingshot-1L (SSH1L) phospho-regulation antibody sampler kit can be used to detect the level of phosphorylation of SSH1L (Ser-978) relative to the total expression level of SSH1L. The kit also includes peptides for antibody blocking experiments and a secondary reagent for antibody detection. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms