Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLINGSHOT BIOSCIENCES StimCytes™ CD20, Lyophilized, 3-mimic kit, 1 kit
SDP

Catalog No. NC3981137
Encompass_Preferred

StimCytes™ CD20 are precision engineered synthetic cell mimics designed to support CAR‑T potency assessment through targeted CD20 specific activation, cytokine release and assay optimization.

Specifications:
Catalog Number: SL-00043
Format: Lyophilized, 3-mimic kit
Package Size: 1 kit
Storage Conditions: -20°C
Shipping Conditions: Wet Ice
Shelf Life: 12 months
Country of Origin: United States
For Research Use Only.

Catalog No. NC3981137 Supplier SLINGSHOT BIOSCIENCES Supplier No. SL00043
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

StimCytes™ CD20 are precision engineered synthetic cell mimics designed to support CAR‑T potency assessment through targeted CD20 specific activation, cytokine release and assay optimization.

Specifications:
Catalog Number: SL-00043
Format: Lyophilized, 3-mimic kit
Package Size: 1 kit
Storage Conditions: -20°C
Shipping Conditions: Wet Ice
Shelf Life: 12 months
Country of Origin: United States
For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.