Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Slingshot inhibitor D3 | 1715076-35-9 | 99.5% | 461.55 g·mol⁻¹ | C25H19NO4S2 | 10MM 1ML
SDP

Catalog No. 5000235435
Encompass_Preferred

Slingshot inhibitor D3 is a potent, selective, reversible, and competitive inhibitor of Slingshot phosphatases (SSH family) used in biochemical and cell-based assays to probe SSH function. It is supplied as a ready-to-use 10 mM solution in DMSO and is intended for inhibition studies, mechanistic assays, and compound screening.

  • Potent inhibition of Slingshot 1 (IC50 = 3 μM) and activity against Slingshot 2 (Ki = 3.9 μM).
  • Supplied as a 10 mM solution in DMSO, 1 mL vial ready for use.
  • High purity (~99.5%) suitable for research applications.
  • Molecular weight 461.55 g·mol⁻¹ and formula C25H19NO4S2.
  • Useful for biochemical and cell-based assays to study Slingshot-mediated dephosphorylation pathways.

Catalog No. 50-002-35435 Supplier Medchemexpress LLC Supplier No. HY12436610MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Slingshot inhibitor D3 is a potent, selective, reversible, and competitive inhibitor of Slingshot phosphatases (SSH family) used in biochemical and cell-based assays to probe SSH function. It is supplied as a ready-to-use 10 mM solution in DMSO and is intended for inhibition studies, mechanistic assays, and compound screening.

  • Potent inhibition of Slingshot 1 (IC50 = 3 μM) and activity against Slingshot 2 (Ki = 3.9 μM).
  • Supplied as a 10 mM solution in DMSO, 1 mL vial ready for use.
  • High purity (~99.5%) suitable for research applications.
  • Molecular weight 461.55 g·mol⁻¹ and formula C25H19NO4S2.
  • Useful for biochemical and cell-based assays to study Slingshot-mediated dephosphorylation pathways.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.