Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Safety-Walk™ Slip Resistant General Purpose Tape, Series 600

Catalog No. 17623928 Shop All 3M Products
Change view
Click to view available options
Color:
Black
Width (Metric):
25 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Width (Metric)
17-623-928 Black 25 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-623-928 Supplier 3M™ Safety-Walk™ Supplier No. 7100383426
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mineral coated, slip resistant material features a highly durable surface. Low profile design helps reduce tripping hazards.

  • Anti-slip tape for light to heavy shoe-traffic areas
  • High quality abrasive particles to ensure long-lasting friction in any application
  • Highly durable surface withstands the rigors of equipment traffic
  • High performance, pressure-sensitive adhesive for easy application
  • Custom shapes and sizes are also available
  • NFSI High Traction Certification
TRUSTED_SUSTAINABILITY

Specifications

Finish Mineral-coated
Width (English) 1 in.
Color Black
Dimensions (L x W) 18.3 m x 25 mm
For Use With (Application) Light to Heavy Traffic Areas
Length (English) 60 ft.
Length (Metric) 18.3 m
Packaging Quantity 4 Pk.
Width (Metric) 25 mm
Adhesive Type Pressure Sensitive Adhesive
Type Floor Tape
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.