Learn More
Grainger Slip Tee Connector 3/4 in CTS PEX White
Catalog No. 502764031
Encompass_Preferred
The John Guest Slip Tee Connector is a plastic push-fit plumbing fitting for use with polyethylene tubing. It provides a leak-free connection without requiring clamps or tools. This 1/4" slip tee connector is made of acetal plastic for durability and has a maximum pressure rating of 232 PSI at 73°F. It is NSF/ANSI 61 certified for potable water applications. The slip tee connector allows a branch line to be added to the main tubing line. It is ideal for use in water filtration, icemakers, humidifiers, RO systems and more. John Guest fittings utilize an o-ring seal design for a secure connection.
Catalog No. 50-276-4031 Supplier Grainger Supplier No. 16T729
The John Guest Slip Tee Connector is a plastic push-fit plumbing fitting for use with polyethylene tubing. It provides a leak-free connection without requiring clamps or tools. This 1/4" slip tee connector is made of acetal plastic for durability and has a maximum pressure rating of 232 PSI at 73°F. It is NSF/ANSI 61 certified for potable water applications. The slip tee connector allows a branch line to be added to the main tubing line. It is ideal for use in water filtration, icemakers, humidifiers, RO systems and more. John Guest fittings utilize an o-ring seal design for a secure connection.