Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FyterTech Nonwovens Slipfyter™ Traction Mat

Catalog No. 19066671 Shop All NPS Corporation Products
Change view
Click to view available options
Capacity (English):
10.5 gal.
21 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-066-671 10.5 gal.
19-066-670 21 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-066-671 Supplier FyterTech Nonwovens Supplier No. STM9800GRY
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Slipfyter™ Traction Mats roll out easily for continuous coverage at entrances and walkways. No gaps, ripples or overlaps that can be slip, trip or fall hazards

  • Easy, durable solution to combat slips, trips and falls in a variety of locations
  • Withstand heavy foot and vehicle traffic, including fork trucks
  • Bottom layer sticks to the ground/surface with Cling technology
  • Peel up and re-position as needed
  • Durable top layer can be vacuumed, swept with broom or mopped
  • ADA compliant for wheelchairs and walkers
  • A fraction of the weight of rubber-backed mats: easing back strain when changing mats
  • Create custom-sized rugs for transition areas and problem spots where leaks or spills may be present

Specifications

Color Gray
Description Traction Rug
Capacity (English) 10.5 gal.
Capacity (Metric) 40 L
Material Needle-punch Polypropylene
Type Absorbent Rug
For Use With (Application) Floors, walkways, entryways
Packaging Quantity 1 Roll
Weight Capacity Medium-weight
Dimensions (L x W x H) 10 x 10 x 34.375 in.
Length (English) 10 in.
Length (Metric) 0.254 m
Width (English) 10 in.
Width (Metric) 0.254 m
Height (English) 34.375 in.
Height (Metric) 0.87 m
Diameter (English) 10 in.
Diameter (Metric) 0.254 m
Weight (English) 15 lb.
Weight (Metric) 7 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.