Learn More
Research Products International Corp Sliplock Flipper Rack, Four Racks (Green, Yellow, Red and Blue) and Two Lids per Case
SDP
Catalog No. 501367543
Encompass
Sliplock Flipper Racks feature an innovative design that provides an expandable workstation. Use racks individually or interlock up to four modules for multiple tube storage.
Racks accommodate up to
  • 24 x 1.5 or 2.0ml tubes
  • 16 x 0.5ml tubes
  • 6 x 8 tube strips
  • 5 x 12 tubes strips
  • one 96-well plate.
Constructed of durable polypropylene with excellent chemical resistance and can also be autoclaved. Packaged four racks (Green, Yellow, Red and Blue) and two clear lids per case.
Catalog No. 50-136-7543 Supplier Research Products International Corp Supplier No. 218405
Sliplock Flipper Racks feature an innovative design that provides an expandable workstation. Use racks individually or interlock up to four modules for multiple tube storage.
Racks accommodate up to
  • 24 x 1.5 or 2.0ml tubes
  • 16 x 0.5ml tubes
  • 6 x 8 tube strips
  • 5 x 12 tubes strips
  • one 96-well plate.
Constructed of durable polypropylene with excellent chemical resistance and can also be autoclaved. Packaged four racks (Green, Yellow, Red and Blue) and two clear lids per case.