Learn More
Electron Microscopy Sciences Slip-On Cover 50- Microscope Slide Box
SDP
Catalog No. 50931103
Encompass_Preferred
Microscope slide box with slip on cover is made from durable polystyrene (with heavy walls). This material is unaffected by humidity and is thoroughly insect proof. The cork lining prevents damage to the stored slides. To ensure proper slide identification, each slot is numbered to correspond to the slide inventory sheet on the inside cover of the box. To save space the boxes are stackable. Each box stores up to 50 3x1" or 25x75 mm microscope slides.
Catalog No. 50-931-103 Supplier Electron Microscopy Sciences Supplier No. 71462
Microscope slide box with slip on cover is made from durable polystyrene (with heavy walls). This material is unaffected by humidity and is thoroughly insect proof. The cork lining prevents damage to the stored slides. To ensure proper slide identification, each slot is numbered to correspond to the slide inventory sheet on the inside cover of the box. To save space the boxes are stackable. Each box stores up to 50 3x1" or 25x75 mm microscope slides.