Learn More
ABclonal Technology SLIRP Rabbit pAb
SDP
Catalog No. 5000105835
Encompass_Preferred
Steroid receptor RNA activator (SRA, or SRA1, MIM 603819) is a complex RNA molecule containing multiple stable stem-loop structures that functions in coactivation of nuclear receptors. SLIRP interacts with stem-loop structure-7 of SRA (STR7) and modulates nuclear receptor transactivation (Hatchell et al., 2006 [PubMed 16762838]).
Catalog No. 50-001-05835 Supplier ABclonal Technology Supplier No. A15524500UL
Steroid receptor RNA activator (SRA, or SRA1, MIM 603819) is a complex RNA molecule containing multiple stable stem-loop structures that functions in coactivation of nuclear receptors. SLIRP interacts with stem-loop structure-7 of SRA (STR7) and modulates nuclear receptor transactivation (Hatchell et al., 2006 [PubMed 16762838]).