Learn More
Aviva Systems Biology Corp SLIT1 Antibody - middle region (ARP58765_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58765_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLIT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SLIT1. Alternate names: MEGF4, SLIL1, SLIT3, Slit-1, SLIT-1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62416