Learn More
Aviva Systems Biology Corp SLIT3 Antibody - N-terminal region (ARP58766_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58766_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SLIT3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SLIT3. Alternate names: FLJ10764, MEGF5, SLIL2, SLIT1, Slit-3, slit2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62417