Learn More
Carnabio Usa Inc SLK/10UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 0712910
Encompass
SLK, Full-length, Active mutant [S5N], Amino Acid 1-1152, NP_055535.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0987