Learn More
Medchemexpress LLC Slk/stk10-in-1 | 264210-49-3 | 99.8% | C17H13ClN2O3 | 10MG
SDP
Catalog No. 5000261494
Encompass_Preferred
SLK/STK10-IN-1 is a potent, selective small-molecule inhibitor of the serine/threonine kinases SLK and STK10 with reported antitumor activity. Supplied as a solid for research use, it combines high purity and defined physicochemical properties for biochemical and cellular studies.
  • Potent, selective inhibitor of SLK and STK10 with reported antitumor activity.
  • High purity suitable for research applications (99.8%).
  • Supplied as a solid with molecular weight 328.75 and formula C17H13ClN2O3.
  • Available in small milligram scales for laboratory experiments.
  • Stable under recommended storage conditions (powder stored at -20°C for long-term stability).
Catalog No. 50-002-61494 Supplier Medchemexpress LLC Supplier No. HY13286810MG
SLK/STK10-IN-1 is a potent, selective small-molecule inhibitor of the serine/threonine kinases SLK and STK10 with reported antitumor activity. Supplied as a solid for research use, it combines high purity and defined physicochemical properties for biochemical and cellular studies.
  • Potent, selective inhibitor of SLK and STK10 with reported antitumor activity.
  • High purity suitable for research applications (99.8%).
  • Supplied as a solid with molecular weight 328.75 and formula C17H13ClN2O3.
  • Available in small milligram scales for laboratory experiments.
  • Stable under recommended storage conditions (powder stored at -20°C for long-term stability).