Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Slm6031434 hydrochloride | 1897379-34-8 | 99.1% | 1 MG
SDP

Catalog No. 5000234471
Encompass_Preferred

A selective sphingosine kinase 2 (SphK2) inhibitor supplied as the hydrochloride salt for research use. Solid, high-purity material characterized by molecular formula C22H31ClF3N5O2 and molecular weight 489.96 Da.

  • Selective sphingosine kinase 2 inhibitor for biochemical and cellular studies.
  • Hydrochloride salt form, supplied as a solid powder.
  • High purity (reported 99.1%), suitable for research applications.
  • Soluble in water at approximately 40 mg/mL with ultrasonic warming if required.
  • Recommended storage: long-term in solvent at -80°C, short-term at -20°C, sealed and away from moisture.

Catalog No. 50-002-34471 Supplier Medchemexpress LLC Supplier No. HY120268A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A selective sphingosine kinase 2 (SphK2) inhibitor supplied as the hydrochloride salt for research use. Solid, high-purity material characterized by molecular formula C22H31ClF3N5O2 and molecular weight 489.96 Da.

  • Selective sphingosine kinase 2 inhibitor for biochemical and cellular studies.
  • Hydrochloride salt form, supplied as a solid powder.
  • High purity (reported 99.1%), suitable for research applications.
  • Soluble in water at approximately 40 mg/mL with ultrasonic warming if required.
  • Recommended storage: long-term in solvent at -80°C, short-term at -20°C, sealed and away from moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.