Learn More
Cayman Chemical SLM6031434 (hydrochloride), Purity: ≥98%, CAS: 1897379-34-8, 10 mg
SDP
Catalog No. 502991017
Encompass_Preferred
An SPHK2 inhibitor (Ki = 0.4 μM for the recombinant mouse enzyme); selective for SPHK2 over SPHK1 (Kis = >20 μM); decreases S1P and increases sphingosine levels in U937 monocytic leukemia cells in a concentration-dependent manner; reduces blood S1P levels in Sphk1-/-, but not Sphk2-/-, mice at 5 mg/kg; increases blood S1P levels in wild-type mice and rats
Catalog No. 50-299-1017 Supplier Cayman Chemical Supplier No. 3043410 MG
An SPHK2 inhibitor (Ki = 0.4 μM for the recombinant mouse enzyme); selective for SPHK2 over SPHK1 (Kis = >20 μM); decreases S1P and increases sphingosine levels in U937 monocytic leukemia cells in a concentration-dependent manner; reduces blood S1P levels in Sphk1-/-, but not Sphk2-/-, mice at 5 mg/kg; increases blood S1P levels in wild-type mice and rats