Learn More
Electron Microscopy Sciences Veco Slot Grid with Handle HD L2x1, Nickel with 2000x1000 µm Slot
SDP
Catalog No. 5026435
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter and 0.8 mil thickness.
Material: Nickel
Slot: 2000 x 1000 µm
Catalog No. 50-264-35 Supplier Electron Microscopy Sciences Supplier No. HD2010NI
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter and 0.8 mil thickness.
Material: Nickel
Slot: 2000 x 1000 µm