Learn More
Electron Microscopy Sciences Gilder Single Slot Oval Grid G102, Gold, 1000 x 200 µm Slot
SDP
Catalog No. 501901268
Encompass_Preferred
Single Slot TEM Grids with oval holes have a thickness of are 50 microns. Each grid is individually inspected.
Material: Gold
Slot: 1000 x 200 µm
Catalog No. 50-190-1268 Supplier Electron Microscopy Sciences Supplier No. G102AU
Single Slot TEM Grids with oval holes have a thickness of are 50 microns. Each grid is individually inspected.
Material: Gold
Slot: 1000 x 200 µm