Learn More
Electron Microscopy Sciences Gilder Single Slot Oval Grid G102, Copper, 1000 x 200 µm Slot
SDP
Catalog No. 501901266
Encompass_Preferred
Single Slot TEM Grids with oval holes have a thickness of are 50 microns. Each grid is individually inspected.
Material: Copper
Slot: 1000 x 200 µm
Catalog No. 50-190-1266 Supplier Electron Microscopy Sciences Supplier No. G102CU
Single Slot TEM Grids with oval holes have a thickness of are 50 microns. Each grid is individually inspected.
Material: Copper
Slot: 1000 x 200 µm