Learn More
Electron Microscopy Sciences Gilder Single Slot Oval Grid G153, Copper, 1500 x 300 µm Slot
SDP
Catalog No. 501901269
Encompass_Preferred
Single Slot TEM Grids with oval holes have a thickness of are 50 microns. Each grid is individually inspected.
Material: Copper
Slot: 1500 x 300 µm
Catalog No. 50-190-1269 Supplier Electron Microscopy Sciences Supplier No. G153CU
Single Slot TEM Grids with oval holes have a thickness of are 50 microns. Each grid is individually inspected.
Material: Copper
Slot: 1500 x 300 µm